siRNA Details
| Gene Symbol: ITGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181501.1 | NP_852478.1 | 25, 26 | 3231 | ||
| GenBank mRNA | AK300423.1 | - | 25, 26 | 3396 | ||
| BC137121.1 | - | 25, 26 | 3230 | 9021118 | ||
| BC137122.1 | - | 25, 26 | 3230 | 9021119 | ||
| BX640981.1 | - | 14, 15 | 5854 | |||
| BX648284.1 | - | 3, 4 | 3059 | |||
| X68742.1 | - | 24, 25 | 3095 | |||

ITGA1
Genomic Map