siRNA Details
| Gene Symbol: ITGAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002208.4 | NP_002199.3 | 22, 23 | 2845 | ||
| GenBank mRNA | BC113436.1 | - | 22, 23 | 2835 | 8322488 | |
| BC117207.1 | - | 22, 23 | 2835 | 40125758 | ||
| L25851.2 | - | 22, 23 | 2871 | |||

ITGAE
Genomic Map