siRNA Details
| Gene Symbol: ITIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002216.2 | NP_002207.2 | 5, 6 | 633 | ||
| GenBank mRNA | AK225329.1 | - | 5, 6 | 538 | ||
| AK291379.1 | - | 5, 6 | 581 | |||
| BC132685.1 | - | 5, 6 | 520 | 40146707 | ||
| X07173.1 | - | 5, 6 | 534 | |||

ITIH2
Genomic Map