siRNA Details
| Gene Symbol: JARID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267040.1 | NP_001253969.1 | 4 | 496 | ||
| NM_004973.3 | NP_004964.2 | 4 | 996 | |||
| XM_005249089.3 | XP_005249146.1 | 3 | 334 | |||
| XM_011514579.2 | XP_011512881.1 | 6 | 1140 | |||
| XM_011514584.2 | XP_011512886.1 | 2 | 250 | |||
| XM_017010833.1 | XP_016866322.1 | 6 | 1140 | |||
| XM_017010834.1 | XP_016866323.1 | 3 | 854 | |||
| XM_017010835.1 | XP_016866324.1 | 2 | 184 | |||
| GenBank mRNA | AK292861.1 | - | 4 | 842 | ||
| AK299349.1 | - | 3 | 410 | |||
| AK303610.1 | - | 4 | 439 | |||
| BC046246.1 | - | 4 | 713 | 4520786 | ||
| U57592.1 | - | 4 | 675 | |||

JARID2
Genomic Map