siRNA Details
| Gene Symbol: KCNC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004977.2 | NP_004968.2 | 2 | 1920 | ||
| XM_006723203.2 | XP_006723266.1 | 2 | 1625 | |||
| XM_011526925.2 | XP_011525227.1 | 2 | 1625 | |||
| XM_011526926.1 | XP_011525228.1 | 2 | 1625 | |||
| XM_011526928.2 | XP_011525230.1 | 2 | 1625 | |||
| GenBank mRNA | AB208930.1 | - | 1 | 726 | ||
| AF055989.1 | - | 2 | 1920 | |||

KCNC3
Genomic Map