siRNA Details
| Gene Symbol: KCNJ13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172416.1 | NP_001165887.1 | 2 | 150 | ||
| NM_002242.4 | NP_002233.2 | 2 | 150 | |||
| GenBank mRNA | AF061118.1 | - | 2 | 101 | ||
| AJ006128.1 | - | 1 | 56 | |||
| AJ007557.1 | - | 2 | 129 | |||
| AK314019.1 | - | 2 | 150 | |||
| AY758240.1 | - | 1 | 32 | |||
| AY758241.1 | - | 1 | 32 | |||
| BC037290.2 | - | 2 | 150 | 4817527 | ||
| DA278958.1 | - | 2 | 148 | |||
| DB173405.1 | - | 2 | 150 | |||

KCNJ13
Genomic Map