siRNA Details
| Gene Symbol: KIF2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098511.2 | NP_001091981.1 | 7 | 926 | ||
| NM_001243952.1 | NP_001230881.1 | 8 | 1129 | |||
| NM_001243953.1 | NP_001230882.1 | 7 | 869 | |||
| NM_004520.4 | NP_004511.2 | 7 | 926 | |||
| GenBank mRNA | AK297347.1 | - | 7 | 672 | ||
| AK302270.1 | - | 7 | 563 | |||
| AK307880.1 | - | 7 | 580 | |||
| AK315866.1 | - | 7 | 553 | |||
| AY317140.1 | - | 5 | 457 | |||
| BC031828.2 | - | 6 | 551 | 4932771 | ||
| EF560716.1 | - | 7 | 637 | |||
| EF560728.1 | - | 7 | 631 | |||
| Y08319.1 | - | 6 | 552 | |||

KIF2A
Genomic Map