siRNA Details
| Gene Symbol: KLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002258.2 | NP_002249.1 | 2 | 174 | ||
| GenBank mRNA | AK292022.1 | - | 2 | 189 | ||
| BC113997.1 | - | 2 | 162 | |||
| BC114516.1 | - | 2 | 162 | 40031215 | ||
| BF432813.1 | - | 2 | 175 | |||
| U11276.1 | - | 2 | 172 | |||

KLRB1
Genomic Map