siRNA Details
| Gene Symbol: KPNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002267.3 | NP_002258.2 | 15 | 1638 | ||
| XM_017020561.1 | XP_016876050.1 | 15 | 1321 | |||
| GenBank mRNA | AF005263.1 | - | 15 | 1235 | ||
| AF034756.1 | - | 15 | 1225 | |||
| AK223480.1 | - | 15 | 1260 | |||
| AK290528.1 | - | 15 | 1298 | |||
| AK291000.1 | - | 15 | 1262 | |||
| BC017355.2 | - | 15 | 1247 | 5020312 | ||
| BC024202.2 | - | 15 | 1247 | 3531069 | ||
| D89618.1 | - | 15 | 1314 | |||
| Y12394.1 | - | 15 | 1238 | |||

KPNA3
Genomic Map