siRNA Details
| Gene Symbol: KRT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000223.3 | NP_000214.1 | 1 | 386 | ||
| GenBank mRNA | AK313747.1 | - | 1 | 400 | ||
| D78367.1 | - | 1 | 386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT12| Gene Symbol: KRT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000223.3 | NP_000214.1 | 1 | 386 | ||
| GenBank mRNA | AK313747.1 | - | 1 | 400 | ||
| D78367.1 | - | 1 | 386 | |||