siRNA Details
| Gene Symbol: KRT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000422.2 | NP_000413.1 | 8 | 1342 | ||
| GenBank mRNA | AK095342.1 | - | 8 | 1271 | ||
| BC000159.2 | - | 8 | 1271 | 2986100 | ||
| BC011901.2 | - | 8 | 1269 | 4111703 | ||
| BC056421.1 | - | 8 | 1271 | 5208304 | ||
| BC072018.1 | - | 9 | 1209 | 3531173 | ||
| BC072019.1 | - | 8 | 1267 | 5442690 | ||
| BX647923.1 | - | 7 | 1608 | |||
| X05803.1 | - | 7 | 723 | |||
| X62571.1 | - | 8 | 1269 | |||

KRT17
Genomic Map