siRNA Details
| Gene Symbol: LCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005565.3 | NP_005556.1 | 21 | 1772 | ||
| GenBank mRNA | AK122637.1 | - | 2 | 1049 | ||
| AK292890.1 | - | 21 | 1733 | |||
| AK296098.1 | - | 13 | 1129 | |||
| AK300000.1 | - | 19 | 1616 | |||
| BC016618.1 | - | 21 | 1709 | 4476274 | ||
| BT007273.1 | - | 21 | 1565 | |||
| FN432830.1 | - | 22 | 1645 | |||
| U20158.1 | - | 21 | 1820 | |||

LCP2
Genomic Map