siRNA Details
| Gene Symbol: LPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167671.2 | NP_001161143.1 | 5 | 551 | ||
| NM_001167672.2 | NP_001161144.1 | 4 | 520 | |||
| NM_005578.4 | NP_005569.1 | 5 | 608 | |||
| XM_005247446.4 | XP_005247503.1 | 5 | 585 | |||
| XM_005247450.4 | XP_005247507.1 | 4 | 554 | |||
| XM_005247451.4 | XP_005247508.1 | 5 | 585 | |||
| XM_005247453.2 | XP_005247510.1 | 4 | 557 | |||
| XM_011512820.2 | XP_011511122.1 | 7 | 852 | |||
| XM_011512823.2 | XP_011511125.1 | 4 | 2667 | |||
| XM_011512827.2 | XP_011511129.1 | 5 | 508 | |||
| XM_011512828.2 | XP_011511130.1 | 5 | 673 | |||
| XM_011512831.2 | XP_011511133.1 | 4 | 415 | |||
| XM_011512833.2 | XP_011511135.1 | 6 | 642 | |||
| XM_011512834.2 | XP_011511136.1 | 6 | 733 | |||
| XM_017006377.1 | XP_016861866.1 | 7 | 714 | |||
| XM_017006378.1 | XP_016861867.1 | 6 | 730 | |||
| XM_017006379.1 | XP_016861868.1 | 6 | 865 | |||
| XM_017006380.1 | XP_016861869.1 | 6 | 761 | |||
| XM_017006381.1 | XP_016861870.1 | 5 | 551 | |||
| XM_017006382.1 | XP_016861871.1 | 3 | 432 | |||
| GenBank mRNA | AK304035.1 | - | 4 | 462 | ||
| AL833171.1 | - | 5 | 779 | |||
| BC130584.1 | - | 4 | 410 | 40146615 | ||
| BC144076.1 | - | 5 | 467 | 9052596 | ||
| CD697098.1 | - | 4 | 540 | |||
| CR457074.1 | - | 3 | 362 | |||
| U49957.1 | - | 5 | 608 | |||

LPP
Genomic Map