siRNA Details
| Gene Symbol: CYP4F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000896.2 | NP_000887.2 | 3 | 371 | ||
| XM_011528014.2 | XP_011526316.1 | 1 | 123 | |||
| GenBank mRNA | AB002454.1 | - | 3 | 371 | ||
| AK298675.1 | - | 3 | 351 | |||
| D12620.1 | - | 3 | 355 | |||
| D12621.1 | - | 3 | 337 | |||

CYP4F3
Genomic Map