siRNA Details
| Gene Symbol: LUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002345.3 | NP_002336.1 | 3 | 1295 | ||
| GenBank mRNA | AK223177.1 | - | 3 | 980 | ||
| AK312682.1 | - | 3 | 998 | |||
| BC007038.1 | - | 3 | 1047 | 3950745 | ||
| BC035997.1 | - | 3 | 969 | 4688606 | ||
| BT006707.1 | - | 2 | 906 | |||
| U18728.1 | - | 3 | 986 | |||
| U21128.1 | - | 3 | 990 | |||

LUM
Genomic Map