siRNA Details
| Gene Symbol: MAB21L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005584.4 | NP_005575.1 | 1 | 1517 | ||
| GenBank mRNA | AB073388.1 | - | 1 | 1507 | ||
| AB593021.1 | - | 1 | 1438 | |||
| AB593022.1 | - | 1 | 1508 | |||
| AK313185.1 | - | 1 | 1493 | |||
| BC028170.1 | - | 1 | 1438 | 5244028 | ||
| CR457420.1 | - | 1 | 960 | |||

MAB21L1
Genomic Map