siRNA Details
| Gene Symbol: MAGEA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011543.2 | NP_001011543.2 | 5 | 540 | ||
| NM_001251828.1 | NP_001238757.1 | 5 | 468 | |||
| NM_021048.4 | NP_066386.2 | 4 | 449 | |||
| GenBank mRNA | AK314161.1 | - | 4 | 449 | ||
| BC004105.2 | - | 4 | 424 | 3926146 | ||
| CB158974.1 | - | 5 | 508 | |||
| CB959556.1 | - | 5 | 551 | |||
| DA827990.1 | - | 5 | 430 | |||

MAGEA10
Genomic Map