siRNA Details
| Gene Symbol: MCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085377.1 | NP_001078846.1 | 10 | 1998 | ||
| NM_002387.2 | NP_002378.1 | 8 | 1542 | |||
| XM_005271991.2 | XP_005272048.1 | 8 | 1576 | |||
| XM_017009473.1 | XP_016864962.1 | 10 | 1597 | |||
| XM_017009474.1 | XP_016864963.1 | 8 | 4262 | |||
| GenBank mRNA | AK096212.1 | - | 7 | 1144 | ||
| AK128596.1 | - | 10 | 2010 | |||
| BC009279.2 | - | 8 | 1542 | 4111706 | ||
| BC018919.2 | - | 8 | 1542 | 4131685 | ||
| BX537952.1 | - | 8 | 1013 | |||
| BX647710.1 | - | 10 | 1472 | |||
| M62397.1 | - | 8 | 1232 | |||

MCC
Genomic Map