siRNA Details
| Gene Symbol: MFAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005926.2 | NP_005917.2 | 6 | 968 | ||
| GenBank mRNA | AK312867.1 | - | 6 | 944 | ||
| BC023557.2 | - | 6 | 903 | 4139958 | ||
| BC050742.1 | - | 6 | 914 | 6200464 | ||
| U04209.1 | - | 6 | 902 | |||

MFAP1
Genomic Map