siRNA Details
| Gene Symbol: MKI67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145966.1 | NP_001139438.1 | 12 | 1751 | ||
| NM_002417.4 | NP_002408.3 | 13 | 2831 | |||
| XM_006717864.3 | XP_006717927.2 | 2 | 732 | |||
| XM_011539818.2 | XP_011538120.1 | 10 | 2562 | |||
| GenBank mRNA | X65550.1 | - | 13 | 2832 | ||
| X65551.1 | - | 12 | 1752 | |||

MKI67
Genomic Map