siRNA Details
| Gene Symbol: MAP3K10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002446.3 | NP_002437.2 | 5, 6 | 1727 | ||
| XM_006723222.2 | XP_006723285.1 | 3, 4 | 586 | |||
| XM_006723223.3 | XP_006723286.1 | 2, 3 | 392 | |||
| XM_011526981.2 | XP_011525283.1 | 5, 6 | 1838 | |||
| XM_011526982.2 | XP_011525284.1 | 5, 6 | 1837 | |||
| GenBank mRNA | X90846.1 | - | 5, 6 | 1727 | ||
| Z48615.1 | - | 5, 6 | 1596 | |||

MAP3K10
Genomic Map