siRNA Details
| Gene Symbol: MMP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002428.3 | NP_002419.1 | 8 | 2105 | ||
| GenBank mRNA | BC006412.1 | - | 8 | 1225 | ||
| BC036495.1 | - | 8 | 2111 | 5263145 | ||
| BC055428.1 | - | 8 | 2106 | 4811572 | ||
| BT006715.1 | - | 6 | 1005 | |||
| D85510.1 | - | 5 | 835 | |||
| D86331.1 | - | 7 | 1043 | |||
| Z48482.1 | - | 8 | 1368 | |||

MMP15
Genomic Map