siRNA Details
| Gene Symbol: MMP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002428.3 | NP_002419.1 | 10 | 2390 | ||
| GenBank mRNA | BC006412.1 | - | 10 | 1510 | ||
| BC036495.1 | - | 10 | 2396 | 5263145 | ||
| BC055428.1 | - | 10 | 2391 | 4811572 | ||
| BT006715.1 | - | 8 | 1290 | |||
| D85510.1 | - | 7 | 1120 | |||
| D86331.1 | - | 9 | 1328 | |||
| Z48482.1 | - | 10 | 1653 | |||

MMP15
Genomic Map