siRNA Details
| Gene Symbol: MOCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001075098.3 | NP_001068566.1 | 11 | 2022 | ||
| NM_005943.5 | NP_005934.2 | 10 | 2026 | |||
| NR_033233.1 | 9 | 1832 | ||||
| XM_011514632.2 | XP_011512934.1 | 11 | 1918 | |||
| XM_011514633.2 | XP_011512935.1 | 10 | 1870 | |||
| XM_011514634.2 | XP_011512936.1 | 10 | 1815 | |||
| XM_017010888.1 | XP_016866377.1 | 10 | 1840 | |||
| GenBank mRNA | AF034374.1 | - | 11 | 1905 | ||
| AJ224328.1 | - | 10 | 2023 | |||
| AK056740.1 | - | 9 | 1778 | |||
| AK091306.1 | - | 10 | 1754 | |||
| AY423726.1 | - | 1 | 718 | |||

MOCS1
Genomic Map