siRNA Details
| Gene Symbol: MYBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278610.1 | NP_001265539.1 | 12 | 2054 | ||
| NM_002466.3 | NP_002457.1 | 13 | 2126 | |||
| GenBank mRNA | AK223482.1 | - | 13 | 1861 | ||
| AK303249.1 | - | 12 | 1959 | |||
| AK314791.1 | - | 13 | 2031 | |||
| BC007585.1 | - | 13 | 2051 | 3162656 | ||
| BC053555.1 | - | 13 | 2020 | 6140320 | ||
| BX647151.1 | - | 3 | 2511 | |||
| X13293.1 | - | 13 | 1988 | |||

MYBL2
Genomic Map