siRNA Details
| Gene Symbol: MYBPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004997.2 | NP_004988.2 | 7, 8 | 1158 | ||
| GenBank mRNA | BC044226.1 | - | 7, 8 | 1158 | 6176021 | |
| L05606.1 | - | 7, 8 | 1127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYBPH| Gene Symbol: MYBPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004997.2 | NP_004988.2 | 7, 8 | 1158 | ||
| GenBank mRNA | BC044226.1 | - | 7, 8 | 1158 | 6176021 | |
| L05606.1 | - | 7, 8 | 1127 | |||