siRNA Details
| Gene Symbol: MYH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017533.2 | NP_060003.2 | 31 | 4429 | ||
| XM_017024676.1 | XP_016880165.1 | 29 | 4573 | |||
| GenBank mRNA | AF111783.2 | - | 31 | 4429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYH4| Gene Symbol: MYH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017533.2 | NP_060003.2 | 31 | 4429 | ||
| XM_017024676.1 | XP_016880165.1 | 29 | 4573 | |||
| GenBank mRNA | AF111783.2 | - | 31 | 4429 | ||