siRNA Details
| Gene Symbol: MYO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012334.2 | NP_036466.2 | 33 | 4995 | ||
| XM_005248306.4 | XP_005248363.1 | 15 | 3889 | |||
| XM_005248307.2 | XP_005248364.1 | 15 | 3365 | |||
| XM_006714475.2 | XP_006714538.1 | 32 | 4702 | |||
| XM_011514046.2 | XP_011512348.1 | 15 | 3068 | |||
| GenBank mRNA | AB018342.2 | - | 33 | 4982 | ||
| AF132021.1 | - | 33 | 4607 | |||
| AF234532.1 | - | 33 | 4749 | |||
| AF247457.2 | - | 33 | 4680 | |||
| AK074717.1 | - | 9 | 1341 | |||
| AK295630.1 | - | 8 | 1243 | |||
| AK295736.1 | - | 15 | 2850 | |||
| AK303459.1 | - | 3 | 589 | |||
| AK316138.1 | - | 15 | 3023 | |||
| AL832428.1 | - | 23 | 3449 | |||
| BC137168.1 | - | 33 | 4704 | 9021165 | ||
| BC150285.1 | - | 33 | 4982 | 8860110 | ||

MYO10
Genomic Map