siRNA Details
| Gene Symbol: NAP1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021963.3 | NP_068798.1 | 1 | 1457 | ||
| GenBank mRNA | AB027013.1 | - | 1 | 1431 | ||
| AF136178.1 | - | 1 | 1405 | |||
| AK291297.1 | - | 1 | 1415 | |||
| AK303676.1 | - | 2 | 1050 | |||
| AK316569.1 | - | 1 | 1457 | |||
| BC026325.1 | - | 1 | 1379 | 4798964 | ||
| CR457095.1 | - | 1 | 1075 | |||

NAP1L2
Genomic Map