siRNA Details
| Gene Symbol: NDUFA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002490.3 | NP_002481.2 | 3 | 443 | ||
| GenBank mRNA | AF047182.1 | - | 3 | 307 | ||
| AK291874.1 | - | 3 | 321 | |||
| AK316562.1 | - | 3 | 317 | |||
| BC002772.1 | - | 3 | 304 | 3619356 | ||
| CR456529.1 | - | 3 | 443 | |||
| CR456714.1 | - | 3 | 303 | |||
| CR541997.1 | - | 3 | 303 | |||

NDUFA6
Genomic Map