siRNA Details
| Gene Symbol: NDUFB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199958.1 | NP_001186887.1 | 2 | 196 | ||
| NM_002492.3 | NP_002483.1 | 2 | 196 | |||
| GenBank mRNA | AF047181.1 | - | 2 | 173 | ||
| AK313822.1 | - | 2 | 174 | |||
| AW955118.1 | - | 2 | 153 | |||
| BC005271.1 | - | 2 | 187 | 3997377 | ||
| BC009796.1 | - | 2 | 191 | 4284776 | ||
| BC017909.1 | - | 2 | 186 | |||
| BG776403.1 | - | 2 | 202 | |||
| DA112751.1 | - | 2 | 199 | |||

NDUFB5
Genomic Map