siRNA Details
| Gene Symbol: NDUFV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021074.4 | NP_066552.2 | 8 | 837 | ||
| XM_017025782.1 | XP_016881271.1 | 8 | 1027 | |||
| GenBank mRNA | AF147367.1 | - | 4 | 312 | ||
| AK312129.1 | - | 8 | 755 | |||
| AW051350.1 | - | 1 | 97 | |||
| BC001632.1 | - | 8 | 747 | 3536345 | ||
| BC017487.1 | - | 8 | 745 | 3537815 | ||
| BC071689.1 | - | 8 | 727 | 4372795 | ||
| CR456928.1 | - | 8 | 723 | |||
| M22538.1 | - | 8 | 741 | |||

NDUFV2
Genomic Map