siRNA Details
| Gene Symbol: NHS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136024.3 | NP_001129496.1 | 7 | 1806 | ||
| NM_001291867.1 | NP_001278796.1 | 7 | 2401 | |||
| NM_001291868.1 | NP_001278797.1 | 6 | 1743 | |||
| NM_198270.3 | NP_938011.1 | 6 | 2338 | |||
| XM_011545528.2 | XP_011543830.1 | 4 | 1950 | |||
| GenBank mRNA | AK302437.1 | - | 7 | 2587 | ||
| AY436752.1 | - | 6 | 2338 | |||
| AY456992.1 | - | 7 | 1806 | |||
| AY456993.1 | - | 6 | 2461 | |||
| BC136415.1 | - | 7 | 2127 | 9020403 | ||
| BC171763.1 | - | 7 | 2127 | 9054416 | ||
| CR936788.1 | - | 6 | 1512 | |||
| GQ988776.1 | - | 7 | 2066 | |||

NHS
Genomic Map