siRNA Details
Gene Symbol: NKX2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002509.3 | NP_002500.1 | 2 | 1086 | ||
XM_006723566.3 | XP_006723629.1 | 2 | 743 | |||
GenBank mRNA | BC075092.2 | - | 2 | 757 | 30915603 | |
BC075093.2 | - | 2 | 757 | 30915391 |
Gene Symbol: NKX2-2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI675189.1 | - | 2 | 342 |
