siRNA Details
| Gene Symbol: NOTCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200001.1 | NP_001186930.1 | 18 | 3248 | ||
| NM_024408.3 | NP_077719.2 | 18 | 3248 | |||
| XM_005270901.3 | XP_005270958.1 | 19 | 5562 | |||
| XM_011541519.2 | XP_011539821.1 | 18 | 8356 | |||
| XM_011541520.2 | XP_011539822.1 | 18 | 5266 | |||
| XM_017001372.1 | XP_016856861.1 | 18 | 2918 | |||
| XM_017001373.1 | XP_016856862.1 | 18 | 2897 | |||
| GenBank mRNA | AF308601.1 | - | 18 | 2963 | ||
| AF315356.1 | - | 18 | 2963 | |||
| BC071562.1 | - | 18 | 3197 | 4822955 | ||
| U77493.1 | - | 1 | 53 | |||

NOTCH2
Genomic Map