siRNA Details
| Gene Symbol: NOTCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000435.2 | NP_000426.2 | 31, 32 | 5891 | ||
| XM_005259924.4 | XP_005259981.1 | 30, 31 | 5674 | |||
| GenBank mRNA | U97669.1 | - | 31, 32 | 5893 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOTCH3| Gene Symbol: NOTCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000435.2 | NP_000426.2 | 31, 32 | 5891 | ||
| XM_005259924.4 | XP_005259981.1 | 30, 31 | 5674 | |||
| GenBank mRNA | U97669.1 | - | 31, 32 | 5893 | ||