siRNA Details
| Gene Symbol: NPAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321307.1 | NP_001308236.1 | 17 | 4270 | ||
| NM_002519.2 | NP_002510.2 | 17 | 4249 | |||
| XM_011542854.2 | XP_011541156.1 | 17 | 4276 | |||
| XM_017017802.1 | XP_016873291.1 | 17 | 5014 | |||
| XM_017017803.1 | XP_016873292.1 | 17 | 4457 | |||
| GenBank mRNA | AK290020.1 | - | 17 | 4249 | ||
| AK301256.1 | - | 4 | 1520 | |||
| BC136270.1 | - | 17 | 4170 | 9020257 | ||
| D83243.1 | - | 17 | 4213 | |||
| X97186.1 | - | 17 | 4181 | |||

NPAT
Genomic Map