siRNA Details
| Gene Symbol: NPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000908.3 | NP_000899.1 | 5 | 1697 | ||
| NM_001204375.1 | NP_001191304.1 | 5 | 1697 | |||
| NM_001204376.1 | NP_001191305.1 | 5 | 630 | |||
| XM_005248309.1 | XP_005248366.1 | 5 | 1089 | |||
| XM_011514047.1 | XP_011512349.1 | 5 | 594 | |||
| XM_011514048.2 | XP_011512350.1 | 5 | 577 | |||
| XM_011514049.2 | XP_011512351.1 | 5 | 657 | |||
| XM_017009492.1 | XP_016864981.1 | 4 | 1584 | |||
| GenBank mRNA | AF025998.1 | - | 5 | 1252 | ||
| AK300094.1 | - | 5 | 613 | |||
| AK308301.1 | - | 5 | 613 | |||
| BC131540.1 | - | 5 | 1405 | 40085188 | ||
| M59305.1 | - | 5 | 1614 | |||
| X52282.1 | - | 5 | 1306 | |||

NPR3
Genomic Map