siRNA Details
| Gene Symbol: NTHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318193.1 | NP_001305122.1 | 2 | 374 | ||
| NM_002528.6 | NP_002519.1 | 2 | 374 | |||
| XM_017023253.1 | XP_016878742.1 | 2 | 377 | |||
| GenBank mRNA | AB001575.1 | - | 2 | 338 | ||
| BC000391.2 | - | 2 | 331 | |||
| BC003014.1 | - | 2 | 331 | 2821314 | ||
| BE900738.1 | - | 2 | 256 | |||
| HY057336.1 | - | 2 | 351 | |||
| U79718.1 | - | 2 | 371 | |||
| U81285.1 | - | 2 | 336 | |||
| Y09687.1 | - | 2 | 325 | |||

NTHL1
Genomic Map