siRNA Details
| Gene Symbol: OAZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301020.1 | NP_001287949.1 | 5 | 927 | ||
| NM_004152.3 | NP_004143.1 | 5 | 933 | |||
| GenBank mRNA | AL542477.3 | - | 3 | 504 | ||
| D78361.1 | - | 5 | 964 | |||
| D87914.1 | - | 5 | 880 | |||
| U09202.1 | - | 5 | 865 | |||

OAZ1
Genomic Map