siRNA Details
| Gene Symbol: OR3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002551.3 | NP_002542.3 | 1 | 967 | ||
| XM_017024697.1 | XP_016880186.1 | 6 | 1587 | |||
| XM_017024698.1 | XP_016880187.1 | 4 | 1279 | |||
| GenBank mRNA | BC095517.1 | - | 1 | 967 | 6971961 | |

OR3A2
Genomic Map