siRNA Details
| Gene Symbol: OXCT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000436.3 | NP_000427.1 | 5 | 836 | ||
| XM_017009494.1 | XP_016864983.1 | 5 | 717 | |||
| GenBank mRNA | AK312327.1 | - | 5 | 532 | ||
| BC009001.1 | - | 5 | 560 | 4148643 | ||
| CR457049.1 | - | 5 | 505 | |||
| EF095213.1 | - | 2 | 242 | |||
| U62961.1 | - | 5 | 603 | |||

OXCT1
Genomic Map