siRNA Details
| Gene Symbol: PAX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006193.2 | NP_006184.2 | 1, 2 | 316 | ||
| XM_011516276.2 | XP_011514578.1 | 4, 5 | 590 | |||
| GenBank mRNA | AB008913.1 | - | 1, 2 | 134 | ||
| AF043978.1 | - | 1, 2 | 316 | |||
| BC074761.2 | - | 1, 2 | 143 | 30915220 | ||

PAX4
Genomic Map