siRNA Details
| Gene Symbol: PDE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003683.2 | NP_001003683.1 | 5 | 640 | ||
| NM_001258312.1 | NP_001245241.1 | 5 | 667 | |||
| NM_001258313.1 | NP_001245242.1 | 4 | 819 | |||
| NM_001258314.1 | NP_001245243.1 | 3 | 496 | |||
| NM_005019.4 | NP_005010.2 | 4 | 682 | |||
| XM_011511323.2 | XP_011509625.1 | 4 | 459 | |||
| XM_011511324.2 | XP_011509626.1 | 5 | 687 | |||
| XM_011511325.2 | XP_011509627.1 | 4 | 814 | |||
| XM_011511326.2 | XP_011509628.1 | 3 | 491 | |||
| XM_017004294.1 | XP_016859783.1 | 4 | 682 | |||
| XM_017004295.1 | XP_016859784.1 | 5 | 687 | |||
| XM_017004296.1 | XP_016859785.1 | 5 | 609 | |||
| XM_017004297.1 | XP_016859786.1 | 5 | 550 | |||
| XM_017004298.1 | XP_016859787.1 | 5 | 732 | |||
| XM_017004299.1 | XP_016859788.1 | 5 | 687 | |||
| XM_017004300.1 | XP_016859789.1 | 5 | 732 | |||
| XM_017004301.1 | XP_016859790.1 | 5 | 537 | |||
| XM_017004302.1 | XP_016859791.1 | 4 | 814 | |||
| GenBank mRNA | AJ401610.1 | - | 4 | 401 | ||
| AK294239.1 | - | 5 | 602 | |||
| AK295657.1 | - | 5 | 472 | |||
| AK301720.1 | - | 3 | 479 | |||
| AL110263.1 | - | 4 | 793 | |||
| BC022480.1 | - | 4 | 650 | 4814064 | ||
| DC396228.1 | - | 3 | 496 | |||
| U40370.1 | - | 5 | 533 | |||

PDE1A
Genomic Map