siRNA Details
| Gene Symbol: PEX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000286.2 | NP_000277.1 | 2 | 1144 | ||
| GenBank mRNA | AB004546.1 | - | 2 | 713 | ||
| AK312635.1 | - | 2 | 1057 | |||
| BC015751.1 | - | 2 | 1059 | |||
| BC031085.1 | - | 2 | 1048 | 5296541 | ||
| U91521.1 | - | 2 | 1075 | |||

PEX12
Genomic Map