siRNA Details
| Gene Symbol: PGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304451.1 | NP_001291380.1 | 6, 7 | 719 | ||
| NM_001304452.1 | NP_001291381.1 | 7, 8 | 864 | |||
| NM_002631.3 | NP_002622.2 | 7, 8 | 785 | |||
| GenBank mRNA | AK290404.1 | - | 7, 8 | 710 | ||
| AK296889.1 | - | 8, 9 | 613 | |||
| AK298830.1 | - | 7, 8 | 864 | |||
| AK300952.1 | - | 6, 7 | 622 | |||
| AK304423.1 | - | 6, 7 | 642 | |||
| BC000368.2 | - | 7, 8 | 696 | 2819330 | ||
| U30255.1 | - | 6, 7 | 665 | |||

PGD
Genomic Map