siRNA Details
| Gene Symbol: PHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005392.3 | NP_005383.3 | 14 | 2014 | ||
| XM_005252051.2 | XP_005252108.1 | 14 | 2008 | |||
| XM_006717143.2 | XP_006717206.1 | 14 | 1903 | |||
| GenBank mRNA | AB014562.2 | - | 13 | 1907 | ||
| AK294861.1 | - | 2 | 152 | |||
| AL834263.1 | - | 14 | 1865 | |||
| BC028051.2 | - | 2 | 122 | |||
| BC152437.1 | - | 13 | 1907 | |||

PHF2
Genomic Map