siRNA Details
| Gene Symbol: PHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005392.3 | NP_005383.3 | 15, 16 | 2262 | ||
| XM_005252051.2 | XP_005252108.1 | 15, 16 | 2256 | |||
| XM_006717143.2 | XP_006717206.1 | 15, 16 | 2151 | |||
| GenBank mRNA | AB014562.2 | - | 14, 15 | 2155 | ||
| AK294861.1 | - | 3, 4 | 400 | |||
| AL834263.1 | - | 15, 16 | 2113 | |||
| BC028051.2 | - | 3, 4 | 370 | |||
| BC152437.1 | - | 14, 15 | 2155 | |||

PHF2
Genomic Map