siRNA Details
| Gene Symbol: PI4KA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058004.3 | NP_477352.3 | 39 | 4665 | ||
| XM_005261634.1 | XP_005261691.1 | 38 | 4572 | |||
| XM_017028829.1 | XP_016884318.1 | 38 | 4599 | |||
| XM_017028830.1 | XP_016884319.1 | 25 | 2821 | |||
| GenBank mRNA | AB210002.1 | - | 39 | 4635 | ||
| AF012872.1 | - | 38 | 4404 | |||
| AK302424.1 | - | 39 | 4593 | |||
| AK314781.1 | - | 11 | 1319 | |||
| BC018120.1 | - | 9 | 982 | |||
| BC053654.1 | - | 8 | 958 | |||
| L36151.1 | - | 7 | 951 | |||

PI4KA
Genomic Map