siRNA Details
| Gene Symbol: PLIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145311.1 | NP_001138783.1 | 5 | 457 | ||
| NM_002666.4 | NP_002657.3 | 5 | 528 | |||
| XM_005254934.4 | XP_005254991.1 | 5 | 386 | |||
| GenBank mRNA | AB005293.1 | - | 5 | 471 | ||
| BC031084.1 | - | 5 | 471 | 5284402 | ||

PLIN1
Genomic Map